Would you like to log in again?
DrumQuik PRO Series Drum and Tote, Dip Tube and Connection Technology, Colder Products Company
Supplier: CPC (Colder Products Company)
DQPRO120208206 DQPRO20G2NPSST DQPRODT0410 DQPRODT0985 DQPRO202BUT202 DQPRO2056X4202 DQPRO202BUT DQPRO120208 DQPRO2056X4 DQPRODT1070
89097-468PK 441 USD InStock
89097-468 89097-466 89066-568 89097-464 89097-462 89097-460 89097-472 89066-570 89066-566 89066-564
DrumQuik PRO Series Drum and Tote, Dip Tube and Connection Technology, Colder Products Company
Fittings and Connectors
Engineered to provide safe, secure container change outs with a quick connect and disconnect technology and integrated dip tube
Materials of construction include polypropylene or PVDF, HDPE, 316 stainless steel or an all plastic spring, EPDM, FKM or FFKM o-ring seals.
Convert and open dispensing system into a closed dispensing system with the use of any common pump.
Ordering information: Accessories not included
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty