Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

K-9 Beef Treats™, Certified, Bio-Serv

Supplier: Bio-Serv
K-9 Beef Treats™
F05411 F05411
89067-254PK 210.69 USD InStock
89067-254 CA89067-254
K-9 Beef Treats™, Certified, Bio-Serv
Animal Supplies Animal Feed
Nutritionally fortified 5 g grain-based treat is highly palatable and a great appetite stimulant.

  • Hearty roast beef flavor
  • Nutritionally assayed
  • Shelf Life: 6 months, cool dry storage

Ingredients: Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (preserved with BHA), Dried Brewers Yeast, Mineral Mix, Vitamin Mix, Beef Flavor, Magnesium Stearate, Calcium Silicate DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin.

Caloric value: 16.9 kcal/Wafer
Nutritional ash content: 7.9%
Nutritional carbohydrate content: 54.0%
Nutritional fat content: 3.8%
Nutritional fibre content: 3.9%
Nutritional moisture content: <10%
Nutritional protein content: 21.9%

Certifications: Certified (Contaminant Screened).

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty