Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Crumble Disk Treats, Bio-Serv

Supplier: Bio-Serv
F05048CD F05048CD F05048CD-25 F05048CD-25
89067-298PK 224.71 USD InStock
89067-298 CA89067-298
Crumble Disk Treats, Bio-Serv
Animal Supplies Animal Feed
Compressed mixed fruit–flavored foraging crumbles are a nutritionally complete treat with Vitamin C.

  • Treats provide natural foraging and picking behaviors
  • Can be used with several Bio-Serv devices
  • Uncolored Banana Flavor is Certified (Contaminant Screened) and suitable for use in GLP and TOX studies
  • Nutritionally assayed
  • Shelf Life: 6 months, cool dry storage

Ingredients: Dextrose, Casein, Cellulose, Corn Starch, Sucrose, Mineral Mix, Canola Oil, Maltodextrin, Tablet Binder, Choline Dihydrogen Citrate, Vitamin Mix, Ascorbic Acid, DL-Methionine, BHA, Natural and Artificial Flavors, Acesulfame Potassium, Citric Acid, Food Dye (Mixed Fruit Flavor Only).

Caloric value: 116.90 kcal/Block
Nutritional ash content: 4.5%
Nutritional carbohydrate content: 54.0%
Nutritional fat content: 5.0%
Nutritional fibre content: 11.6%
Nutritional moisture content: <10%
Nutritional protein content: 18.3%
Vitamin C: 155 mg/Block
Vitamin D3: 71.2 IU/Block

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty