Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Grainola Trail Mix, Bio-Serv

Supplier: Bio-Serv
F75711 F75711
89067-314EA 193.27 USD InStock
89067-314 CA89067-314
Grainola Trail Mix, Bio-Serv
Animal Supplies Animal Feed
Human food–grade natural cereal foraging mixture.

  • Use with Tuff Turf Foraging Board or scatter into bedding
  • Shelf Life: 6 months, cool dry storage

Ingredients: Rolled Oats, Rolled Wheat, Brown Sugar, Canola Oil, Whey Powder, Coconut, Honey, Almond Flavor, and Salt.

Caloric value: 4.22 kcal/g
Nutritional ash content: 2.0%
Nutritional carbohydrate content: 51.8%
Nutritional fat content: 18.2%
Nutritional fibre content: 7.3%
Nutritional moisture content:<10%
Nutritional protein content: 12.7%

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty