Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nutty Monkey Treats™, Bio-Serv

Supplier: Bio-Serv
Nutty Monkey Treats™
F3788 F3788
89067-324PK 152.54 USD InStock
89067-324 CA89067-324
Nutty Monkey Treats™, Bio-Serv
Animal Supplies Animal Feed
Nutritionally complete grain-based diet with bits of peanuts

  • Excellent source of Vitamin C
  • Nutritionally complete treat or sole source diet
  • No messy shells to sweep or clog drains
  • Nutritionally assayed
  • Shelf Life: 6 months, cool dry storage

Ingredients: Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (preserved with BHA), Peanuts, Dried Brewers Yeast, Mineral Mix, Vitamin Mix, DL-Methionine, Choline Chloride, Calcium Silicate, Magnesium Stearate, Sodium Propionate, Ascorbic Acid, Kaolin, tBHQ.

Caloric value: 18.1 kcal/Wafer
Nutritional ash content: 6.8%
Nutritional carbohydrate content: 52.0%
Nutritional fat content: 7.5%
Nutritional fibre content: 4.5%
Nutritional moisture content: <10%
Nutritional protein content: 21.5%
Vitamin C: 12.6 mg/Wafer
Vitamin D3: 36.9 IU/Wafer

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty