Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Turf Foraging Crumbles™ Treats, Bio-Serv

Supplier: Bio-Serv
Turf Foraging Crumbles™
F05433 F05437 F05433 F05437
89067-408EA 113.28 USD InStock
CA89067-410 89067-408 CA89067-408 89067-410
Turf Foraging Crumbles™ Treats, Bio-Serv
Animal Supplies Animal Feed
Small, fruit-flavored bits, available in a variety of flavors, are nutritionally complete foraging treats

  • Specially sized for use with Bio-Serv's Turf Foraging Board
  • Scatter into bedding to encourage rooting behavior in swine
  • Saves labor and less expensive than perishable fruit
  • Uncolored Banana and Pina Colada Flavors are Certified (Contaminant Screened) and suitable for use in GLP and TOX studies
  • Nutritionally assayed
  • Shelf Life: 12 months, cool dry storage

Ingredients: Dextrose, Casein, Cellulose, Corn Starch, Sucrose, Mineral Mix, Canola Oil, Maltodextrin, Tablet Binder, Choline Dihydrogen Citrate, Vitamin Mix, Ascorbic Acid, DL-Methionine, BHA, Natural and Artificial Flavors, Acesulfame Potassium, Citric Acid, Food Dye.

Caloric value: 3.34 kcal/g
Nutritional ash content: 4.6%
Nutritional carbohydrate content: 53.5%
Nutritional fat content: 5.1%
Nutritional fibre content: 11.8%
Nutritional moisture content: <10%
Nutritional protein content: 18.5%
Vitamin C: 4.5 mg/g
Vitamin D3: 2.1 mg/g

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty