Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bacon Softies™ Treats, Certified, Bio-Serv

Supplier: Bio-Serv
Bacon Softies™
F3580 S3580-1 S3580-1 F3580-1 F3580 F3580-1
89067-424EA 131.09 USD InStock
89067-424 CA89067-424 89067-426 CA89067-426
Bacon Softies™ Treats, Certified, Bio-Serv
Animal Supplies Animal Feed
Soft, pelleted, nutritionally complete diet for rodents and supplement for canines and ferrets.

  • Highly palatable bacon flavor stimulates appetite
  • Excellent soft diet for recovering or debilitated animals and weanlings
  • Replaces labor intensive, nutrient-diluted mush diets
  • Available sterile
  • Nutritionally assayed
  • Shelf Life: 6 months, refrigerated

Ingredients: Soy Flour, Soybean Oil, Corn Syrup, Sucrose, Casein, Cellulose, Corn Starch, Wheat Bran, Molasses, Mineral Mix, Banana Flakes, Acacia Gum, Bacon Flavor, Glycerin, Vitamin Mix, DL-Methionine, Choline Bitartrate, BHA.

Caloric value: 3.83 kcal/g
Nutritional ash content: 4.2%
Nutritional carbohydrate content: 45.8%
Nutritional fat content: 12.0%
Nutritional fibre content: 6.2%
Nutritional moisture content: <10%
Nutritional protein content: 22.4%

Certifications: Certified (Contaminant Screened).

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty