Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dustless Precision Pellets® Primate Purified Diet, Bio-Serv

Supplier: Bio-Serv
Dustless Precision Pellets®
F0079 F0035 F0035 F0059 F0079 F0022 F0059 F0022
89067-544EA 132.93 USD InStock
89067-544 CA89067-532 89067-534 CA89067-544 89067-536 CA89067-534 CA89067-536 89067-532
Dustless Precision Pellets® Primate Purified Diet, Bio-Serv
Animal Supplies Animal Feed
Precise size and weight pellets generally used in nutritional and behavioral studies.

  • Dustless and fracture free
  • Superior performance in automatic feeders
  • Nutritionally complete, banana-flavored, grain-based formula
  • Available in purified formula
  • Wide range of weights: 20 mg to 1 g
  • Every batch tested for hardness, weight and height
  • Custom flavors and colors available upon request
  • Nutritionally assayed
  • Shelf Life: 12 months, cool dry storage

Ingredients: Casein, Dextrose, Sucrose, Cellulose, Corn Starch, Corm Oil, Corn Syrup, Mineral Mix, Choline Bitartrate, Calcium Silicate, Banana Flavor, Vitamin Mix, Magnesium Stearate, L-Cystine, Ascorbic Acid.

Caloric value: 3.45 kcal/g
Nutritional ash content: 5.1%
Nutritional carbohydrate content: 52.0%
Nutritional fat content: 6.3%
Nutritional fibre content: 11.5%
Nutritional moisture content:<10%
Nutritional protein content: 20.2%

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty