Would you like to log in again?
Crumble Disk Treat Holder, Bio-Serv
Supplier: Bio-Serv
9117
89067-738EA 90.34 USD InStock
89067-738
Crumble Disk Treat Holder, Bio-Serv
Animal Supplies Animal Environmental Enrichment
Made of sturdy, 1/8" clear polycarbonate with a 2" inside diameter and stainless steel end caps
Versatile foraging device; hang horizontally or vertically. Primates forage for treats through slot. Removable end caps are secured by stainless steel bolts and nylon lock nuts. Autoclavable. Cagewasher safe.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty