Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Crawl Balls™ Rodent Enrichment Devices, Certified, Bio-Serv

Supplier: Bio-Serv
Crawl Balls™
K3330 K3329 K3330 K3329
89067-842EA 10.51 USD InStock
89067-842 CA89067-842 89067-844 CA89067-844
Crawl Balls™ Rodent Enrichment Devices, Certified, Bio-Serv
Animal Supplies Animal Environmental Enrichment
Crawl balls with three 6.35 cm (2.5") openings. Made of high-temp polycarbonate.

  • Provides a comfortable hiding place
  • Transparent material allows for ease of visibility
  • Autoclavable
  • Cagewasher safe

Recommended for rats up to 300 g, hamsters and gerbils.

Certifications: Certified (Contaminant Screened).

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty