Would you like to log in again?
Mouse and Rat Tunnels, Bio-Serv
Supplier: Bio-Serv
K3323 K3325 K3322 K3326 K3339 K3324-WT K3487 K3323 K3339 K3325 K3326 K3487 K3324-WT K3322
89067-874EA 7.82 USD InStock
CA76353-758 89067-874 CA89067-872 89067-876 CA89067-874 89067-878 CA89067-876 89067-868 CA89067-878 CA89067-868 76353-758 89067-870 89067-872 CA89067-870
Mouse and Rat Tunnels, Bio-Serv
Animal Supplies Animal Environmental Enrichment
Made of sturdy, 3 mm (¹/₈") thick polycarbonate. Autoclavable, cage washer-safe, and certified (contaminant screened).
Optional, sturdy stainless steel round wire hanger can be used to suspend a tunnel above the cage floor.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty