Would you like to log in again?
TRIlites® 984 CP Tri-Polymer Gloves, Mapa Professionnel AdvanTech
Supplier: Mapa Professionnel
TRIlites®
984926 34984027 984929 34984028
89068-068CS 662.43 USD InStock
89068-068 89068-066 89068-064 89068-070
TRIlites® 984 CP Tri-Polymer Gloves, Mapa Professionnel AdvanTech
Gloves
Unique tri-polymer blend of natural rubber, neoprene, and nitrile provides excellent product protection and optimum tactile sensitivity.
  • Comfort of latex with added strength, tear resistance, and chemical splash protection of nitrile and neoprene
  • Textured fingertips deliver superior grip in both wet and dry applications
  • Extra long, 11.5" length offers extended protection for the wrist and upper forearm
  • Powder-free process eliminates cross-contamination
  • Ambidextrous
Gloves are great for laboratory research, light assembly and sorting, precision parts handling, and light maintenance.
Certifications: Complies with 21CFR Parts 170-199.
Packaging: 100 gloves per bag.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty