Would you like to log in again?
VWR® Braid-Reinforced Silicone Tubing
Supplier: VWR
VWR®
VA7100327-25 VA7100210-25 VA7100405-25 VA7100275-25 VA7100158-25
89068-410EA 1222.6 USD InStock
89068-410 89068-430 89068-428 89068-426 89068-412
VWR® Braid-Reinforced Silicone Tubing
Tubing Transfer Tubing
This low-volatile grade, platinum-cured silicone tubing is produced in a certified ISO Class 6 (FED-STD-209E Class 1000/M4.5) cleanroom for critical pharmaceutical, biomedical, food, and cosmetic applications.
  • Sterilizable by autoclave, gamma radiation, SIP, and CIP processes
  • Enhanced performance under pressure without compromising flexibility
The use of polyester yarn braiding in the tubing wall enhances performance capabilities in elevated pressure applications without compromising flexibility
Tubing has undergone extensive physical, chemical, and biological testing and meets USP Class VI, FDA CFR 177.2600, ISO 10993, European Pharmacopoeia 3.1.9., and 3-A standards. Interior silicone is manufactured from NSF 51 listed material.
Tubing is resistant to temperature extremes, compression set, chemical attack, ozone, radiation, moisture, and environmental exposure. Certified free of animal-derived ingredients. Imparts no taste or odor to critical stream.
Ordering information: Supplied in 7.6 m (25') lengths. Sealed in polyethylene bags to ensure cleanliness. Quality control and lot traceability identification on bags and boxes. Complete validation package available upon request.
Caution: Note: Not intended for implantation or continuous steam applications.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty