Would you like to log in again?
VWR® Silicone Tubing
Supplier: VWR
VWR®
VA7002580-50 VA7002957-50 VA7002229-50 VA7002034-50 VA7002216-50 VA7002775-50 VA7002593-50 VA7002021-50 VA7001657-50 VA7002762-50 VA7002398-50 VA7001098-50 VA7002203-50 VA7002567-50
89068-454EA 450.92 USD InStock
89068-454 89068-484 89068-474 89068-460 89068-482 89068-494 89068-480 89068-492 89068-468 89068-458 89068-488 89068-478 89068-456 89068-490
VWR® Silicone Tubing
Tubing Transfer Tubing
Low-volatile grade, platinum-cured silicone tubing is produced in a certified ISO Class 6 (FED-STD-209E Class 1000/M4.5) cleanroom for use in critical pharmaceutical, biomedical, laboratory, food, and cosmetic applications.
  • Non-contaminating
  • Imparts no taste or odor to critical stream
  • Chemically resistant
  • Free of animal-derived ingredients
It is also ideally suited for single use/disposable research and development and production processes.
This extremely flexible tubing resists temperature extremes, compression set, chemical attack, ozone, radiation, moisture, and environmental exposure.
Certifications: Tubing has undergone extensive physical, chemical, and biological testing and meets USP Class VI, FDA CFR 177.2600, ISO 10993, European Pharmacopoeia 3.1.9., and 3-A standards. Manufactured from NSF 51 listed material. Sterilizable via autoclave or gamma radiation.
Ordering information: Supplied in 15.2m (50') lengths. Sealed in polyethylene bags to ensure cleanliness. Quality control and lot traceability identification on bags and boxes. Complete validation package available upon request.
Caution: Note: Not intended for implantation, continuous steam applications, or elevated pressure levels.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty