Would you like to log in again?
Crews® Stryke® Safety Goggles, MCR Safety
Supplier: MCR Safety
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Crews® Stryker®
2322AF 2320AF 2312AF 2310AF
89074-178EA 26.27 USD InStock
89074-178 89068-870 89074-176 89074-174
Crews® Stryke® Safety Goggles, MCR Safety
Goggles
Goggles offer reliable protection from chemical splash while providing a secure, ventilated fit.
  • Stylish, smoke-tinted plastic frame
  • Clear polycarbonate lens with Duramass® scratch-resistant coating
  • Exclusive Duramass AF4® anti-fog coating
  • Rubber strap adjusts easily
Certifications: Goggles meet or exceed ANSI Z87.1 safety standards.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty