Would you like to log in again?
Sterile SatPax® 670 Presaturated Nonwoven Cleanroom Wiper, Berkshire
Supplier: Berkshire
SatPax®
SSP670.00124
89068-952CS 822.53 USD InStock
89068-952
Sterile SatPax® 670 Presaturated Nonwoven Cleanroom Wiper, Berkshire
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Wipers are made from a hydroentangled nonwoven blend of 55% cellulose and 45% polyester wiper.
  • Low extractables and fiber and particle counts
  • High abrasion resistance
  • Reduces alcohol usage and preparation/handling costs
  • Reduces VOC emissions
  • Increases cleaning efficiency
  • Smooth and durable with good wet strength
  • Increases cleaning protocol consistency
Solvent is 70% isopropyl alcohol and 30% deionized water. No chemical binders in base material. Suitable for ISO Class 5 (FED-STD-209E Class 100/M3.5) and less critical environments. Excellent for wet cleaning of critical surfaces where control of flammable solvents and flammable solvent concentrations is required. The 55% cellulose fiber is biodegradeable.
Certifications: Gamma irradiated to a Sterility Assurance Level of 10⁻⁶. Certificate of sterility. Passes LAL test for endotoxins <20 EU/device. Complies with RoHS and REACH requirements. Meets cGMP requirements for traceability.
Packaging: C-fold wipers are packaged in resealable, solvent-resistant packaging.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty