Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Poly Bags, Flat
Supplier: VWR
VWR®
1270 1235 1285 1300 255 625 1000 1210 8135 238 1313 1296 2982 540 1215 1260 1265 55 521 1281 656
89070-448CS 383.28 USD InStock
89070-448 89070-504 89070-438 89070-472 89070-396 89070-374 89069-100 89069-772 89070-372 89070-454 89070-430 89070-442 89069-326 89071-554 89069-678 89069-832 89069-306 89072-360 89070-480 89070-170 89069-644
VWR® Poly Bags, Flat
Bags Storage Bags
Flat bags are made with an extra-strong bottom seal or a durable side weld.
  • Outstanding vapor and moisture barrier
  • Ideal for product presentation
  • Manufactured with 100% renewable energy
Engineered to provide over three times the impact and twice the tensile strength of traditional low-density polyethylene bags.
Certifications: Meet FDA and USDA specifications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty