Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Layflat Poly Bags
Supplier: VWR
VWR®
435 652 142 1280 1135 2235 966 1305 190 1295 535 2415 1275 1201 2358 1142 137 75 402 480 195 170 633 770 875 1137 885 623 1220 106 646 1079 380 1120 15 450 1140 2390 1307 1205 950 1123 112 1186 470 2760 1030 2425 8650 1320 825 2430 925 915 550 1085 970 2800 8635 1010 2350 610 1290 1324 1035 1240
89071-492CS 59.95 USD InStock
89071-492 89071-296 89070-126 89070-368 89070-446 89071-290 89070-406 89070-242 89070-286 89069-122 89069-160 89070-488 89070-444 89070-202 89070-200 89071-456 89069-768 89069-488 89070-084 89069-686 89069-204 89071-240 89069-826 89071-124 89070-252 89069-232 89070-492 89069-596 89070-130 89069-150 89070-376 89070-292 89070-370 89072-482 89069-752 89069-556 89069-814 89072-476 89070-104 89070-308 89070-462 89069-068 89069-262 89069-788 89069-986 89069-668 89069-542 89069-500 89070-180 89071-264 89070-316 89070-514 89070-038 89070-314 89070-518 89069-252 89069-210 89070-470 89070-350 89070-312 89070-034 89069-614 89071-228 89071-306 89070-070 89069-930
VWR® Layflat Poly Bags
Bags Storage Bags
Each of VWR® imported layflat poly bags are made of 100% virgin low density polyethylene (LDPE) resin that meets FDA/USDA specifications.
  • LDPE fabrication
  • Extra strong bottom
  • Superior clarity of siding
Manufactured with an extra strong bottom and a durable side weld for optimal strength. This premium bag features superior clarity for showing the quality of your customer's products. All listed gauges are nominal. 100 Packed. Can be heat sealed, tied, stapled or taped shut.
Certifications: Meet FDA/USDA specifications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty