Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Poly Bags, Gussetted
Supplier: VWR
VWR®
1580 1470 1425 1555 1710 1398 1605 1400 1730 1770 1465 1530 1747 1764
89070-816CS 355.9 USD InStock
89070-816 89070-618 89070-698 89070-864 89070-834 89070-802 89070-714 89070-582 89070-620 89070-686 89070-584 89070-672 89070-596 89070-860
VWR® Poly Bags, Gussetted
Bags Storage Bags
Each of VWR® imported gusseted poly bags are made of 100% virgin Low Density Polyethylene (LDPE) resin.
  • Tubular construction and durable bottom seal
  • Outstanding vapor and moisture barrel
  • Twice the tensile strength of traditional LDPE bags
  • Made with 100% renewable energy
Side gussets expand easily to fit over bulky or irregular size objects. Features superior clarity for showing the quality of the customer's products. 100 packed. All listed gauges are nominal. Can be heat sealed, tied, stapled or taped shut.
Certifications: Meet FDA/USDA specifications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty