Would you like to log in again?
VWR® Low Density PE Tubing
Supplier: VWR
3005 3050 2109 1960 3030 1967 2045 3025
89071-570CS 271.38 USD InStock
89071-570 89071-582 89070-960 89071-572 89071-562 89071-046 89071-002 89070-964
VWR® Low Density PE Tubing
Tubing Transfer Tubing
The clear poly tubing is made of 100% virgin Low Density Polyethylene (LDPE) resin that meets FDA/USDA specifications.
  • Clear LDPE resin fabrication
  • Even thickness and unrivaled clarity
Tubing is manufactured to exact specification to ensure even wall thickness and unrivaled clarity. Neatly wound on a 7.6 cm (3") core with 2.5 cm (1") diameter core plug. Can be heat sealed, tied, stapled or taped shut.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty