Would you like to log in again?
VWR® Poly Bags, Pre-Opened
Supplier: VWR
VWR®
2715
89071-416CS 175.54 USD InStock
89071-416
VWR® Poly Bags, Pre-Opened
Bags Storage Bags
Pre-opened bags on rolls are for use on automated packing machines.
  • Suitable for use with automated packing machines
  • Tubular construction
  • Durable bottom seal
Bags have a slit in front and a perforated back for automatic loading or fast hand loading of products. Made with a tubular construction and a durable bottom seal. Wound on a 8.9 cm (3¹/₂") core. Core plug has a 2.5 cm (1") diameter hole.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty