Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Reclosable Parts Bags
Supplier: VWR
VWR®
4005 4020 4010
89071-890CS 248.03 USD InStock
89071-890 89071-892 89071-896
VWR® Reclosable Parts Bags
Bags Storage Bags
These bags have been chosen for their ability to rival any reclosable bag at economical prices.
  • Full gauge
  • Exceptional clarity and quality
  • Hanging hole
  • Prepared writing area
These reclosable bags are full gauge and offer exceptional clarity and quality. They feature a large white writing area that accepts either marker or pen and a single hang hole above the zipper closure for a secure hang. These tough bags employ a reinforced side-welded zipper to lock in single smooth motion. Made of 100% virgin Low Density Polyethylene (LDPE) resin.
Certifications: Meet FDA/USDA specifications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty