Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
VWR® Reclosable Pink Anti-Static Bags
Supplier: VWR
VWR®
4045
89071-908CS 499.07 USD InStock
89071-908
VWR® Reclosable Pink Anti-Static Bags
Bags Storage Bags
This premium anti-static bag seals out both dust and moisture while controlling static electricity.
  • Amine-free
  • Anti-static properties
  • Reclosable design
  • Pink tint for easy identification
This bag is constructed with a durable single-track zipper that stands up to repeated use with side seam welding to prevent splitting, is amine-free, won't corrode polycarbonate and offers superior performance in low humidity. With a surface resistivity less than 1×1012 ohms/sq VWR's 4 mm bags. This can also be heat sealed, tied, stapled or taped shut.
Certifications: Meets the full static decay requirements of MIL-PRF-81705E, Type II and EIA-541.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty