Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Slider Top Reclosable Bags
Supplier: Avantor
VWR®
4070 4065 4075 4085 4067 4090 4093
89071-914CS 55.08 USD InStock
89071-914 89071-916 89071-928 89071-918 89071-932 89071-922 89071-934
VWR® Slider Top Reclosable Bags
Bags Storage Bags
These three zip top slider bags are great for applications where quick, positive closure and ease of use are important.
  • Zip top design
  • Quick, positive closure
  • Easy to use
  • Made of 100% virgin low density polyethylene (LDPE) resin
Ideal for use with gloved hands. The slider helps hold the bag open for one handed loading.
Certifications: Meet FDA/USDA specifications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty