Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
VWR® High Density Liner Bags
Supplier: VWR
VWR®
5830 5835 5815 5825 5800
89072-218CS 211.29 USD InStock
89072-218 89072-216 89072-206 89072-212 89072-220
VWR® High Density Liner Bags
Bags Waste Disposal Bags
High Density Polyethylene (HDPE) liners provide extra strength at lighter weight.
  • Provided in heavy-duty dispenser box
  • Available in a range of thicknesses
Provides excellent puncture resistance from sharp objects and heavy items. Comes in convenient heavy-duty dispenser box. Star sealed bottoms help prevent tear-out.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty