Would you like to log in again?
VWR® Flat Poly Bags on Rolls
Supplier: VWR
VWR®
6865 6860 6850 6845 6855
89072-306CS 468.95 USD InStock
89072-306 89072-304 89072-302 89072-310 89072-308
VWR® Flat Poly Bags on Rolls
Bags Storage Bags
Flat bags are made with an extra-strong bottom seal or a durable side weld.
  • Three times the impact and twice the tensile strength of traditional LDPE bags
  • Made with 100% renewable energy
Outstanding vapor and moisture barrier. Ideal for product presentation. Wound on 7.6 cm (3") cores. Core plugs have 2.5 cm (1") Ø holes. Engineered to provide over three times the impact and twice the tensile strength of traditional low-density polyethylene bags.
Certifications: Meet FDA and USDA specifications.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty