Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Rubbermaid® Commercial Invader® Side-Gate Wet-Mop Handle

Supplier: Rubbermaid
Invader®
RCPH146 RCPH136 RCPH126 RCPH146GRE
89073-436EA 84.62 USD InStock
89073-436 89073-442 89073-444 75778-008
Rubbermaid® Commercial Invader® Side-Gate Wet-Mop Handle
Mops
Solvent- and acid-resistant frame enables use with all commercial floor stripping agents.

  • Easily remove and replace mop heads
  • Side gate makes wet mop head connection easy
  • Solvent and acid-resistant frame
  • For use with 1" headband mop heads

Easy-change, side-gate design allows you to quickly slide mop heads on or off. Adjustable thumb wheel enables stress-free, tight and secure clamp down.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty