Would you like to log in again?
HYGEN™ Quick Connect Single-Sided Frame, Rubbermaid® Commercial
Supplier: Rubbermaid
HYGEN™
RCPQ560 RCPQ570
89073-574EA 124.35 USD InStock
89073-574 89073-572
HYGEN™ Quick Connect Single-Sided Frame, Rubbermaid® Commercial
Mops
The Rubbermaid Commercial Products HYGEN Mop Quick-Connect Wet/Dry Single-Sided Frame features a flat profile that slides easily under furniture and equipment.
  • For bare floors
  • Connects to microfiber mop heads with a hook-and-loop fastener that can be replaced when worn
  • Features a nylon swivel joint for flexible maneuvering
  • Corrosion resistance
  • Made with non-magnetic metals, enabling use near magnetically sensitive equipment, such as MRIs
The Rubbermaid HYGEN microfiber mop features a lightweight aluminum with stainless steel construction for improved productivity. The one-step connecting mechanism speeds the mop parts assembly and changeover. The flat frame profile is sanitary and easy to clean and the replaceable hook-and-loop strips are cost efficient and extend product life.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty