Would you like to log in again?
PIG® Absorbent Mat Pads in Dispenser Box, New Pig
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MAT251 MAT240 MAT170 MAT171 MAT139 MAT172 MAT155 MAT154 MAT135
89073-820CS 195.8 USD InStock
89073-820 89073-822 89073-828 56620-556 89073-824 46000-492 89073-814 94001-112 89073-826
PIG® Absorbent Mat Pads in Dispenser Box, New Pig
Spill Control Materials
Eight layers of 100% polypropylene are thermally bonded to make PIG Mat exceptionally strong; won't rip, tear or fray even when saturated.
  • Eight-layer pads and rolls combine grip and strength to reduce slip hazards and keep floors clean
  • Dispenser box makes it easy to take, carry and store mats while protecting from dirt and moisture
Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
Absorbs and retains oils, coolants, solvents and water. Dark gray color hides grime so mat stays on the job longer. Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat while on the job. Can be wrung out and incinerated after use to reduce waste or for fuels blending. Dispenser box makes it easy to take, carry and store your mat while protecting it from dirt and moisture. Pads are ideal for catching drips and soaking up spills. Easy-tear perforations let you take only what you need so you use less mat and save money.
Heavyweight construction delivers greater durability and absorbency for demanding tasks and large-volume cleanups. Mediumweight construction offers great performance for everyday tasks and cleanups.
Highly absorbent, fine-fiber construction won't leave liquids or fiber residue behind.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty