Would you like to log in again?
PIG® Heavy-Duty Maintenance Wipers, New Pig
Supplier: New Pig
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PIG®
WIP325 WIP306 WIP303 WIP304 WIP328 WIP329
89073-974CS 103.2 USD InStock
89073-974 89069-044 89069-046 56620-095 89073-972 89069-042
PIG® Heavy-Duty Maintenance Wipers, New Pig
Wipes Disposable Wipes and Towels Wipes for General Applications
Strong and solvent-resistant for wiping machines and rough parts, but soft enough for cleaning skin or preparing surfaces.
  • Excellent for heavy-duty maintenance jobs like machine wipedowns and degreasing parts
  • Low-lint construction won't shed, pill, or leave behind unwanted fibers when wiping parts
  • Binder-free construction withstands solvents, degreasers, and cleaning agents
Wipers are 1/4 folded and poly wrapped for easy handling and loading. Convenient pop-up box lets you take just one wiper at a time while keeping remaining stock clean and dry. Pop-up Z-box keeps wipers clean, neat and organized for quick access. Economical jumbo rolls dispense easily and offer a lower cost per wiper for high-volume use.
Exceptional abrasion resistance and wet strength that stands up to rough surfaces, jagged edges or metal chips.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty