Would you like to log in again?
Ninja® Max Gloves, MCR Safety
Supplier: MCR Safety
Ninja®
N9676GL N9676GXL N9676GM N9676GS
89074-254EA 72.16 USD InStock
89074-254 89074-248 89074-252 89074-250
Ninja® Max Gloves, MCR Safety
Gloves
MCR Safety Ninja® gloves are engineered to provide the highest level of innovation.
  • Made with DSM certified Dyneema®
  • Mild non-ionic detergent should be used
  • 5 to 10 minutes cycle time should be employed
  • Hang to dry, tumble drying is not receommended
Each of the offerings in the Ninja® series include a unique shell and polymer combination incorporating the latest in hand protection technology. Our Ninja® Glove Line offers the greatest assortment of dexterity, sense of touch, and protection. The Ninja® Max features a 10 gauge gray Dyneema®/Synthetic shell, black bi-polymer coated palm and fingertips. Washing instructions include washing gloves in slightly warm water not exceeding.
Ordering information: These gloves are polybagged by the pair.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty