Would you like to log in again?
Adapter, Bleed Capillary, Ace Glass Incorporated
Supplier: Ace Glass
9328-18 9328-02 9328-04 9328-22
89074-400EA 77.08 USD InStock
89074-400 89074-398 89074-396 89074-394
Adapter, Bleed Capillary, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This bleed capillary adapter is fabricated from borosilicate glass.
  • Top [ST] joint
  • With hose connection
  • Fabricated from borosilicate glass
The length of this adapter is measured from the top of the joint to the tip. Used with a 5/16-inch I.D. tubing, size A hose connection.
Ordering information: Contact your Avantor representative for a listing of which flasks and heads fit this adapter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty