Would you like to log in again?
Distillation Column, Vigreux, Microscale, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
9345-32 9345-35
89074-426EA 139.98 USD InStock
89074-426 89074-428
Distillation Column, Vigreux, Microscale, Ace Glass Incorporated
Distillation Components Distillation Columns
Vigreux columm for vacuum distillation of high boiling materials
The top joint is a combination cap, thread and joint to accommodate and seal other microscale apparatus. The O.D. of the top joint has an external thread to facilitate an o-ring and open-top phenolic cap. The internal or I.D. at the top is a ground [ST] 14/10 outer joint. Bottom is a [ST] 14/10 inner joint, and indents. Length stated is length of indents. Fabricated from borosilicate glass.
Ordering information: Supplied with open-top phenolic cap and FETFE o-ring.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty