Would you like to log in again?
Flask, Pear Shaped, Microscale, Single Neck, Ace Glass Incorporated
Supplier: Ace Glass
9477-64 9477-62 9477-60
89074-902EA 50.45 USD InStock
89074-902 89074-900 89074-904
Flask, Pear Shaped, Microscale, Single Neck, Ace Glass Incorporated
Flasks Round Bottom Flasks
Microscale, pear-shaped, single-neck flask
The top joint is a combination cap, thread and joint to accommodate and seal other microscale apparatus. The O.D. of the top joint has an external, ground-in 20-400 thread to facilitate an o-ring and an open-top phenolic cap. The internal (or I.D.) at the top is a ground [ST] 14/10 outer joint. Fabricated from borosilicate glass.
Ordering information: Supplied complete with holed cap, PTFE-faced septum and FETFE o-ring.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty