Would you like to log in again?
Water Cooled Trubore® Bearing, Ace Glass Incorporated
Supplier: Ace Glass
9527-12 9527-14 9527-08
89075-050EA 200.89 USD InStock
89075-050 89075-052 89075-054
Water Cooled Trubore® Bearing, Ace Glass Incorporated
Stirrers Stirring Tools
Jacketed, Trubore® bearing with a standard taper joint for use with ACE precision ground stir shafts
Hose connections fit 5/16" or 3/8" I.D. tubing. Match bearing I.D. of 5 mm or 6 mm to stir shaft O.D. Fabricated from borosilicate glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty