Would you like to log in again?
Micro/Mini-Lab Vacuum Take-Off Adapter with Hose Connection, Ace Glass Incorporated
Supplier: Ace Glass
9564-10
89075-148EA 112.37 USD InStock
89075-148
Micro/Mini-Lab Vacuum Take-Off Adapter with Hose Connection, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
Adapter is used for microscale vacuum distillations.
  • Top joint is a combination cap, thread and [ST] joint
  • Bottom inner joint
  • With hose connection for vacuum
  • 105° angle
  • Cap and O-Ring included
The top joint is a combination cap, thread and standard taper joint to accommodate and seal other microscale apparatus. The O.D. of the top joint has an external thread to facilitate an O-Ring and open-top phenolic cap. The internal or I.D. at the top is a ground standard taper 14/10 outer joint. Inner joint at other end and size C hose connection for vacuum. 105° angle. Supplied with cap and O-Ring.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty