Would you like to log in again?
Micro/Mini-Lab Vacuum Take-Off Adapter with Side Joint, Ace Glass Incorporated
Supplier: Ace Glass
9565-05 9565-06
89075-150EA 106.5 USD InStock
89075-150 89075-152
Micro/Mini-Lab Vacuum Take-Off Adapter with Side Joint, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
Used for Microscale vacuum distillations, especially for use with a spinning band microdistillation column.
  • Top joint is a combination cap, thread and standard taper joint
  • Bottom inner [ST] joint
  • [ST] outer side joint for vacuum connection or stopper
  • 105° angle
  • Cap and O-Ring included
The top joint is a combination cap, thread and standard taper joint to accommodate and seal other microscale apparatus. The O.D. of the top joint has an external thread to facilitate an O-Ring and open-top phenolic cap. The internal or I.D. at the top is a ground outer joint. Inner standard taper joint at other end and a standard taper 7/10 outer side joint for vacuum connection or stopper when using spinning band column technique. 105° angle. Supplied with cap and O-Ring.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty