Would you like to log in again?
Micro/Mini-Lab Stainless Steel Gow-Mac GC Adapter, Ace Glass Incorporated
Supplier: Ace Glass
9571-42 9571-40
89075-172EA 140.22 USD InStock
89075-172 89075-174
Micro/Mini-Lab Stainless Steel Gow-Mac GC Adapter, Ace Glass Incorporated
Fittings and Connectors
Microscale gas chromatography connection-adapter for making direct connection between GC column and 89075-192 collection tube.
  • Stainless steel
  • With NPT female thread and externally threaded 5/5 outer joint
  • Allows “O-Ring- CAP-SEAL” connection via a tube
Stainless steel adapter has a 6-32 NPT female thread for attaching to heated port on GC and a 5/5 outer joint, externally threaded, to allow an “O-Ring- CAP-SEAL” connection via a tube.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty