Would you like to log in again?
Cold Finger Condenser, Microscale, Adjustable, Ace Glass Incorporated
Supplier: Ace Glass
9573-20 9573-08
89075-176EA 21.91 USD InStock
89075-176 89075-178
Cold Finger Condenser, Microscale, Adjustable, Ace Glass Incorporated
Condensers
Micro cold finger condenser with drip tip on the bottom for directing drops into the well of standard taper 14/10 jointed Hickman Stills
Cold water is introduced into cold finger periodically by means of a Pasteur pipet. Complete item consists of cold finger condenser and multi-purpose vacuum adapter with #7 Ace-Thred that makes compression o-ring seal with cold finger and allows for vertical adjustment. Threaded adapter supplied with nylon bushing and FETFE o-ring. Hose barb for use with 5/16-inch I.D. tubing, size A hose connection. Fabricated from borosilicate glass.
Ordering information: Available with cold finger condenser only (89075-176), or complete with cold finger condenser and multi-purpose vacuum adapter with #7 Ace-Thred (89075-178) with bushing and o-ring. For vacuum adapter only, order 89064-824.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty