Would you like to log in again?
Microscale Claisen Adapter, Threaded, Ace Glass Incorporated
Supplier: Ace Glass
9574-24 9574-16 9574-20
89075-180EA 116.04 USD InStock
89075-180 89075-182 89075-184
Microscale Claisen Adapter, Threaded, Ace Glass Incorporated
Distillation Components Distillation Adapters
Microscale Claisen Adapter head is often used to facilitate syringe addition of reagents to sealed or moisture-sensitive systems.
  • Top joints are combination cap, thread and [ST] joints
  • Bottom inner [ST] joint
  • Two threaded caps with hole, two PTFE-faced silicone rubber septa and two o-rings included
The top joints are combination cap, thread and standard taper joints to accommodate and seal other microscale apparatus. The O.D. of the top joints have an external thread to facilitate an o-ring and open-top phenolic cap. The internal or I.D. at the top is a ground outer joint. With inner standard taper joint at bottom. Supplied with two threaded caps with hole, two PTFE-faced silicone rubber septa and two o-rings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty