Would you like to log in again?
Micro Reaction Vials, Conical Bottom, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
9590-20 9590-14 9590-11 9590-04 9590-15 9590-03 9590-19 9590-10 9590-02
89075-232EA 106.93 USD InStock
89075-232 89075-242 89075-234 89075-236 89075-238 89075-226 89075-228 89075-230 89075-240
Micro Reaction Vials, Conical Bottom, Ace Glass Incorporated
Vials
Microscale, heavy-wall reaction vial.
The top joint is a combination cap, thread and joint to accommodate and seal other microscale apparatus. The Ext.Ø. of the top joint has an external thread to facilitate an O-ring and open-top phenolic cap. The internal or Int.Ø. at the top is a ground outer joint. This type connection eliminates the need for clamps while offering the positive leaktight seal of ground glass joints.
Replace O-ring in cap with PTFE-faced silicone rubber septa and vial can be capped for storage.
Delivery information: Each vial supplied with threaded cap with hole, one PTFE faced silicone rubber septum and one O-ring for making 'O-Ring-CAP-SEAL.'
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty