Would you like to log in again?
Microscale Conical Reaction Vials, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
9591-46 9591-23 9591-47 9591-21 9591-48 9591-16
89075-276EA 92.56 USD InStock
89075-276 89075-266 89075-268 89075-270 89075-272 89075-274
Microscale Conical Reaction Vials, Ace Glass Incorporated
Vials
Microscale, thin-wall reaction vial.
The top joint is a combination cap, thread and joint to accommodate and seal other microscale apparatus. The Ext.Ø of the top joint has an external thread to facilitate an O-ring and open-top phenolic cap. The internal or Int.Ø at the top is a ground standard taper 14/10 outer joint. This thin-walled vial has better heat transfer for the more demanding research project. Vials with side ports are supplied with holed cap and septum that facilitates syringe retrieval or injection for the port.
Delivery information: Supplied with threaded cap with hole, one PTFE faced silicone rubber septum and one O-ring. Outer joint for side port vials are also supplied with holed cap, PTFE-faced silicone rubber septum and FETFE O-ring.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty