Would you like to log in again?
Flask, Round Bottom, Microscale, Ace Glass Incorporated
Supplier: Ace Glass
9592-04 9592-15 9592-09 9592-10
89075-278EA 51.56 USD InStock
89075-278 89075-280 89075-282 89075-284
Flask, Round Bottom, Microscale, Ace Glass Incorporated
Flasks Round Bottom Flasks
Microscale, single-neck, round-bottom reaction flask
The top joint is a combination cap, thread and [ST] joint to accommodate and seal other microscale apparatus. The O.D. of the top joint has an external thread to facilitate an o-ring and open-top phenolic cap. The internal or I.D. at the top is a ground outer joint. This type of connection eliminates the need for clamps while offering the positive leak-tight seal of ground glass. Fabricated from borosilicate glass.
Ordering information: Supplied with threaded cap with hole, one PTFE-faced silicone rubber septum, and one o-ring. Part numbers 89075-278, -282 and -284 are designed for use with Cap 89075-248, O-ring 89060-994 and Septa 89064-076. Part number 89075-280 is designed for use with Cap 89075-250, O-ring 89060-962 and Septa 89064-078.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty