Would you like to log in again?
Instatherm® Low-Form Oil Bath, 12-Ft. PTFE-Clad Cord, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
9602-37 9602-13 9602-15 9602-47 9602-41 9602-45 9602-17 9602-43 9602-11 9602-07
89075-364EA 1399.62 USD InStock
89075-364 89075-374 89075-366 89075-376 89075-368 89075-378 89075-360 89075-370 89075-362 89075-372
Instatherm® Low-Form Oil Bath, 12-Ft. PTFE-Clad Cord, Ace Glass Incorporated
Baths Oil Baths
Large, borosilicate glass vessel coated with Ace Instatherm®
Caution: Never run Instatherm® baths dry or breakage can occur, especially at higher temperatures.
Can accept round-bottom flasks up to 5 L, and other large vessels. The bath has a rolled lip for easy handling. The maximum temperature on this bath is 100°C. The full capacity is 15 L without a vessel, and typically should have a minimum of six liters when using a vessel like a 5 L round bottom flask. Baths must be run with a heating media or oil. Three-pronged plug for insertion into Ace temperature controllers.
Ordering information: Ace temperature controller is not included and sold separately; contact your VWR representative for more information. Available as bath alone (with integral 12-foot PTFE-clad controller cord) or complete with clip for sensor and/or thermometer.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty