Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Instatherm® High-Form Oil Bath, Ace Glass Incorporated

Supplier: Ace Glass

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

9603-22 9603-02 9603-04 9603-24 9603-06 9603-20
89075-386EA 1549.59 USD InStock
89075-386 89075-388 89075-380 89075-390 89075-382 89075-384
Instatherm® High-Form Oil Bath, Ace Glass Incorporated
Baths Oil Baths
High form, borosilicate glass open vessels coated with Ace Instatherm®

Three-prong connector plug is designed to work with Ace temperature controllers where the output voltage can be automatically limited to match the bath. Bath can also be matched with ACE temperature thermocouple sensors for complete automatic operation. If using another type temperature controller, voltage should not exceed the rated voltage (stated below).

Caution: Never run Instatherm® baths dry or breakage can occur, especially at higher temperatures.

Accommodates larger and taller vessels. Designed to operate up to 250°C, heating response time is very rapid, (5° per minute), with very low thermal lag. The bottom is uncoated so the bath can be put on any popular magnetic stirrer for stirring while heating. A thermocouple well is imbedded in the insulation in this series. Baths are designed to run with oil or a heating media.

Ordering information: Ace temperature controller is not included and sold separately; contact your Avantor representative for more information. Available as bath alone (with integral six-foot controller cord) or complete with clip for sensor and/or thermometer.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty