Would you like to log in again?
Micro/Mini-Lab Photochemical Reactor with Giant #25 Ace-Thred, Ace Glass Incorporated
Supplier: Ace Glass
9718-39 9718-19 9718-23 9718-12 9718-49
89075-452EA 546.75 USD InStock
89075-452 89075-454 89075-456 89075-458 89075-450
Micro/Mini-Lab Photochemical Reactor with Giant #25 Ace-Thred, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Photochemical
Microscale photochemical reactor with internally threaded connections both top and bottom of head
Top is #15 Ace-Thred that accepts 12.5-14 mm O.D. tubes such as 89075-462 finger condenser; bottom is #25 Ace-Thred that accepts a 24 mm O.D. quartz or glass (borosilicate) reactor tube for photolysis experiments. Complete item consists of head, #15 and #25 nylon bushings with O-Rings, cap stopper with cap and O-Ring, finger condenser, and either quartz or glass 25 x 120 mm reactor tube.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty