Would you like to log in again?
Ultrasonic Booster, Ace Glass Incorporated
Supplier: Ace Glass
9822-20
89075-516EA 1397.71 USD InStock
89075-516
Ultrasonic Booster, Ace Glass Incorporated
Homogenizers Dispersing Tools
When connected between the converter and horn, the booster increases the amplitude of vibration at the horn tip by a factor of two
Use to process very difficult applications
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty