Would you like to log in again?
Ultrasonic Reaction Vessel, Small Volume, 3 to 10 ml, Ace Glass Incorporated
Supplier: Ace Glass
9843-04 9843-25
89075-546EA 441.03 USD InStock
89075-546 89075-548
Ultrasonic Reaction Vessel, Small Volume, 3 to 10 ml, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
Tapered walls and proper size horn allow volumes as little as 3 mL to be mixed
Fabricated of borosilicate glass with #25 Ace-Thred center neck and two standard taper 14/20 side necks. With #25 bushing, center neck will accept #36 slide adapter in 25 mm with a horn and extender. Vessel measures 123 mm (4 7/8") high. Not supplied with bushing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty