Would you like to log in again?
Ultrasonic Jacketed Reaction Vessel, 250 ml, Ace Glass Incorporated
Supplier: Ace Glass
9848-07 9848-35
89075-554EA 1047.41 USD InStock
89075-554 89075-556
Ultrasonic Jacketed Reaction Vessel, 250 ml, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
This vessel is jacketed to provide active cooling from tap water or mechanical cooler
Jacket is cylindrical with flat bottom for greater stability. With #25 Ace-Thred center neck and three standard taper 14/20 side necks. Use #25 bushing with o-ring in center neck to secure #36 slide adapter in 25 mm for connecting converter. Capacity 250 mL. Use with 3/8" I.D. tubing, size D hose connection.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty