Would you like to log in again?
Ultrasonic Jacketed Reaction Vessel, 10-50 mL, Ace Glass Incorporated
Supplier: Ace Glass
9850-12 9850-30
89075-558EA 1034.17 USD InStock
89075-558 89075-560
Ultrasonic Jacketed Reaction Vessel, 10-50 mL, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Flask Shape
This vessel is jacketed to provide active cooling from tap water or mechanical cooler
Jacket is cylindrical with flat bottom for greater stability. With #25 Ace-Thred center neck and two standard taper 14/20 side necks. Use #25 bushing with o-ring in center neck to secure #36 slide adapter in 25 mm for connecting converter. Capacity 10–50 mL. Use with 3/8" I.D. tubing, size D hose connection.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty